1-Benzyl-5-oxopyrrolidine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzyl-5-oxo-3-pyrrolidinecarboxylic Acid

OR6330-25G OR6330-5G
APOSOR6330-25GEA 273 EUR
APOSOR6330-25G APOSOR6330-5G
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid 97%
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219.24 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085749
CAS-Nummer: 5733-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR