1-Methyl-5-oxopyrrolidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR6333-1G OR6333-5G
APOSOR6333-1GEA 92.8 EUR
APOSOR6333-1G APOSOR6333-5G
1-Methyl-5-oxopyrrolidine-3-carboxylic acid 95%
1-Methyl-5-oxopyrrolidine-3-carboxylic acid
Formel: C₆H₉NO₃
Molecular Weight: 143.14 g/mol
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014102
CAS-Nummer: 42346-68-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR