5-Oxo-1-(1,3-thiazol-2-yl)pyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR6337-1G OR6337-250MG OR6337-500MG OR6337-2.5G OR6337-5G
APOSOR6337-1GEA 340 EUR
APOSOR6337-1G APOSOR6337-250MG APOSOR6337-500MG APOSOR6337-2.5G APOSOR6337-5G
5-Oxo-1-(1,3-thiazol-2-yl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(1,3-thiazol-2-yl)pyrrolidine-3-carboxylic acid
Formel: C₈H₈N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD07364777
CAS-Nummer: 914637-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR