1-{[(2-Methyl-2-propanyl)oxy]carbonyl}-3-azetidincarbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid
OR6348-25G
OR6348-100G
APOSOR6348-25GEA
39.3
EUR
APOSOR6348-25G
APOSOR6348-100G
1-{[(2-Methyl-2-propanyl)oxy]carbonyl}-3-azetidincarbonsäure 97%
N-Boc-azetidine-3-carboxylic acid
Formel:
C₉H₁₅NO₄ Molecular Weight: 201,22 g/mol Schmelzpunkt: 98…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860897 CAS-Nummer: 142253-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...