Pyrido[2,3-b]pyrazine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6353-250MG OR6353-500MG
APOSOR6353-250MGEA 286 EUR
APOSOR6353-250MG APOSOR6353-500MG
Pyrido[2,3-b]pyrazine-2-carboxylic acid 97%
Pyrido[2,3-b]pyrazine-2-carboxylic acid
Formel: C₈H₅N₃O₂
Dichte: 1,51 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD08056316
CAS-Nummer: 914637-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR