Imidazo[1,2-a]pyridin-6-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR6355-1G OR6355-100MG OR6355-5G
APOSOR6355-1GEA 24.8 EUR
APOSOR6355-1G APOSOR6355-100MG APOSOR6355-5G
Imidazo[1,2-a]pyridin-6-carbonsäure
Imidazo[1,2-a]pyridin-6-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 244…247 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07021498
CAS-Nummer: 139022-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR