Imidazo[1,2-b]pyridazin-2-carbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR6356-1G OR6356-250MG
APOSOR6356-1GEA 122 EUR
APOSOR6356-1G APOSOR6356-250MG
Imidazo[1,2-b]pyridazin-2-carbonsäure 97%
Imidazo[1,2-b]pyridazin-2-carbonsäure
Formel: C₇H₅N₃O₂
Molecular Weight: 163.1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235229
CAS-Nummer: 160911-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR