Picolinoylchlorid Hydrochlorid 93%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Pyridine-2-carbonyl chloride hydrochloride

OR6370-1G OR6370-5G OR6370-10G
APOSOR6370-1GEA 21.9 EUR
APOSOR6370-1G APOSOR6370-5G APOSOR6370-10G
Picolinoylchlorid Hydrochlorid 93%
Picolinoylchlorid Hydrochlorid
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,0160 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00060193
CAS-Nummer: 39901-94-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR