2,5-Dimethylthiophene-3-carbonyl chloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR6371-1G
OR6371-250MG
APOSOR6371-1GEA
263
EUR
APOSOR6371-1G
APOSOR6371-250MG
2,5-Dimethylthiophene-3-carbonyl chloride 95%
2,5-Dimethylthiophene-3-carbonyl chloride
Formel:
C₇H₇ClOS Lagertemperatur: Kühlschrank |
MDL:
MFCD08056318 CAS-Nummer: 57248-13-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...