4-Methoxythiophene-3-carbonyl chloride

Lieferant: Apollo Scientific

OR6372-1G OR6372-250MG OR6372-500MG
APOSOR6372-1GEA 497 EUR
APOSOR6372-1G APOSOR6372-250MG APOSOR6372-500MG
4-Methoxythiophene-3-carbonyl chloride
4-Methoxythiophene-3-carbonyl chloride
Formel: C₆H₅ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08056319
CAS-Nummer: 224649-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR