2-Oxo-2H-pyran-5-carbonyl chloride

Lieferant: Apollo Scientific

OR6384-1G OR6384-250MG OR6384-500MG OR6384-2.5G
APOSOR6384-1GEA 440 EUR
APOSOR6384-1G APOSOR6384-250MG APOSOR6384-500MG APOSOR6384-2.5G
2-Oxo-2H-pyran-5-carbonyl chloride
2-Oxo-2H-pyran-5-carbonyl chloride
Formel: C₆H₃ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08235241
CAS-Nummer: 23090-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR