Ethyl-5-amino-3-methylisothiazole-4-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6385-250MG
OR6385-1G
OR6385-5G
OR6385-100MG
APOSOR6385-250MGEA
116
EUR
APOSOR6385-250MG
APOSOR6385-1G
APOSOR6385-5G
APOSOR6385-100MG
Ethyl-5-amino-3-methylisothiazole-4-carboxylate 97%
Ethyl-5-amino-3-methylisothiazole-4-carboxylate
Formel:
C₇H₁₀N₂O₂S Molecular Weight: 186,23 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00097916 CAS-Nummer: 34859-65-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...