Methylcumalat 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-oxo-2H-pyran-5-carboxylate
OR6407-100G
OR6407-25G
OR6407-5G
APOSOR6407-100GEA
491
EUR
APOSOR6407-100G
APOSOR6407-25G
APOSOR6407-5G
Methylcumalat 97%
Methylcumalat
Formel:
C₇H₆O₄ Molecular Weight: 154,12 g/mol Siedepunkt: 178…180 °C/60mmHg Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010120 CAS-Nummer: 6018-41-3 EINECS: 227-871-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...