(1-Methyl-1H-imidazol-2-yl)methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-2-imidazolemethanol
OR6426-25G
OR6426-5G
APOSOR6426-25GEA
29
EUR
APOSOR6426-25G
APOSOR6426-5G
(1-Methyl-1H-imidazol-2-yl)methanol 97%
(1-Methyl-1H-imidazol-2-yl)methanol
Formel:
C₅H₈N₂O Molecular Weight: 112.13 g/mol Schmelzpunkt: 107…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00964673 CAS-Nummer: 17334-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...