3-Pyridazinemethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pyridazin-3-ylmethanol, 3-(Hydroxymethyl)pyridazine
OR6428-1G
OR6428-250MG
OR6428-5G
APOSOR6428-1GEA
98.6
EUR
APOSOR6428-1G
APOSOR6428-250MG
APOSOR6428-5G
3-Pyridazinemethanol 97%
3-Pyridazinemethanol
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Siedepunkt: 150 °C/1mmHg Schmelzpunkt: 59…61 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04972629 CAS-Nummer: 37444-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...