4-Methyl-3-nitrophenyl isocyanate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR6434-10G
OR6434-1G
OR6434-5G
APOSOR6434-10GEA
192
EUR
APOSOR6434-10G
APOSOR6434-1G
APOSOR6434-5G
4-Methyl-3-nitrophenyl isocyanate 97%
4-Methyl-3-nitrophenyl isocyanate
When melted, may contain small amount of sediment.
Formel:
C₈H₆N₂O₃ Molecular Weight: 178,15 g/mol Siedepunkt: 113…118 °C/3mmHg Schmelzpunkt: 48…54 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014698 CAS-Nummer: 13471-69-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...