Ethyl-2-(bromomethyl)benzoate 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethyl 2-(bromomethyl)benzoate

OR6441-1G OR6441-5G OR6441-100MG OR6441-250MG
APOSOR6441-1GEA 104 EUR
APOSOR6441-1G APOSOR6441-5G APOSOR6441-100MG APOSOR6441-250MG
Ethyl-2-(bromomethyl)benzoate 97%
Ethyl-2-(bromomethyl)benzoate
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235262
CAS-Nummer: 7115-91-5
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR