4-Acetoxy-3-methoxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Vanillinacetat, 4-Formyl-2-methoxyphenylacetat

OR6464-100G OR6464-1KG OR6464-500G
APOSOR6464-100GEA 58 EUR
APOSOR6464-100G APOSOR6464-1KG APOSOR6464-500G
4-Acetoxy-3-methoxybenzaldehyd
4-Acetoxy-3-methoxybenzaldehyd
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003362
CAS-Nummer: 881-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR