3-(4-Iodophenyl)propionic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Iodophenyl)propanoic acid
OR6478-1G
OR6478-250MG
APOSOR6478-1GEA
52.2
EUR
APOSOR6478-1G
APOSOR6478-250MG
3-(4-Iodophenyl)propionic acid
3-(4-Iodophenyl)propionic acid
Radiopharmaceutical precursor.
Formel:
C₉H₉IO₂ Molecular Weight: 276.08 g/mol Schmelzpunkt: 140…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017597 CAS-Nummer: 1643-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...