4-Amino-1-methylpiperidin 92%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Methylpiperidin-4-amine
OR6512-25G
OR6512-5G
APOSOR6512-25GEA
33.5
EUR
APOSOR6512-25G
APOSOR6512-5G
4-Amino-1-methylpiperidin 92%
4-Amino-1-methylpiperidin
Formel:
C₆H₁₄N₂ Molecular Weight: 114.1888 g/mol Siedepunkt: 83 °C/40mmHg Dichte: 0.91 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD01570549 CAS-Nummer: 41838-46-4 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...