3-Ethoxycarbonylcoumarin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl Coumarin-3-carboxylate, Coumarin-3-carboxylic acid ethyl ester
OR6525-25G
OR6525-1G
OR6525-5G
APOSOR6525-25GEA
226
EUR
APOSOR6525-25G
APOSOR6525-1G
APOSOR6525-5G
3-Ethoxycarbonylcoumarin 98%
3-Ethoxycarbonylcoumarin
Formel:
C₁₂H₁₀O₄ Molecular Weight: 218,21 g/mol Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016964 CAS-Nummer: 1846-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...