3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propansäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

OR6529-25G OR6529-1G OR6529-5G
APOSOR6529-25GEA 101 EUR
APOSOR6529-25G APOSOR6529-1G APOSOR6529-5G
3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propansäure 98%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,196 g/mol
Schmelzpunkt: 149…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023096
CAS-Nummer: 3339-73-9
EINECS: 222-083-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR