3-(4-Hydroxyphenyl)propionsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR6534-100G
OR6534-10G
OR6534-25G
APOSOR6534-100GEA
26.1
EUR
APOSOR6534-100G
APOSOR6534-10G
APOSOR6534-25G
3-(4-Hydroxyphenyl)propionsäure 98%
3-(4-Hydroxyphenyl)propionsäure
Formel:
C₉H₁₀O₃ Schmelzpunkt: 126…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002778 CAS-Nummer: 501-97-3 EINECS: 207-931-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...