1H-Benzimidazol-2-ylmethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Hydroxymethyl)benzimidazole, 2-Benzimidazolemethanol, (2-Benzimidazolyl)methanol
OR6550-10G
OR6550-25G
OR6550-5G
APOSOR6550-10GEA
48
EUR
APOSOR6550-10G
APOSOR6550-25G
APOSOR6550-5G
1H-Benzimidazol-2-ylmethanol 97%
(1H-Benzoimidazol-2-yl)methanol
Formel:
C₈H₈N₂O Molecular Weight: 148.1619 g/mol Schmelzpunkt: 160…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014560 CAS-Nummer: 4856-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...