3,5-Dichlor-1,2,4-thiadiazol 95%

Lieferant: Apollo Scientific
Achtung

OR6558-1G OR6558-5G
APOSOR6558-1GEA 87 EUR
APOSOR6558-1G APOSOR6558-5G
3,5-Dichlor-1,2,4-thiadiazol 95%
3,5-Dichloro-1,2,4-thiadiazole
Formel: C₂Cl₂N₂S
Molecular Weight: 155.01 g/mol
Siedepunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00799286
CAS-Nummer: 2254-88-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR