3-Methoxyzimtsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Methoxyphenyl)acrylic acid

OR6573-100G OR6573-10G OR6573-25G
APOSOR6573-100GEA 162 EUR
APOSOR6573-100G APOSOR6573-10G APOSOR6573-25G
3-Methoxyzimtsäure 97%
3-Methoxyzimtsäure
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Schmelzpunkt: 115…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004386
CAS-Nummer: 6099-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR