4-Methoxyzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Methoxyphenyl)acrylic acid

OR6574-100G OR6574-25G
APOSOR6574-100GEA 52.2 EUR
APOSOR6574-100G APOSOR6574-25G
4-Methoxyzimtsäure 98%
4-Methoxyzimtsäure
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,18 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004398
CAS-Nummer: 830-09-1
EINECS: 212-594-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR