trans-3-Chlorocinnamic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-3-(3-Chlorophenyl)acrylic acid

OR6577-100G OR6577-1KG OR6577-500G
APOSOR6577-100GEA 106 EUR
APOSOR6577-100G APOSOR6577-1KG APOSOR6577-500G
trans-3-Chlorocinnamic acid 98%
trans-3-Chlorzimtsäure
Formel: C₉H₇ClO₂
Molecular Weight: 182.6037 g/mol
Schmelzpunkt: 161…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004384
CAS-Nummer: 14473-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR