3-Nitrozimtsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Nitrophenyl)acrylic acid
OR6582-100G
OR6582-25G
APOSOR6582-100GEA
154
EUR
APOSOR6582-100G
APOSOR6582-25G
3-Nitrozimtsäure 97%
3-Nitrozimtsäure
Formel:
C₉H₇NO₄ Molecular Weight: 193,160 g/mol Schmelzpunkt: 202…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007277 CAS-Nummer: 555-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...