3-[4-(tert-Butyl)phenyl]acrylic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-tert-Butylcinnamic acid
OR6586-10G
OR6586-5G
OR6586-25G
APOSOR6586-10GEA
192
EUR
APOSOR6586-10G
APOSOR6586-5G
APOSOR6586-25G
3-[4-(tert-Butyl)phenyl]acrylic acid 98%
3-[4-(tert-Butyl)phenyl]acrylic acid
Formel:
C₁₃H₁₆O₂ Molecular Weight: 204.27 g/mol Schmelzpunkt: 198…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040719 CAS-Nummer: 1208-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...