2,3-Dimethyl-4-nitroanisol

Lieferant: Apollo Scientific

Synonyme: 1,2-Dimethyl-3-methoxy-6-nitrobenzol, 3-Methoxy-6-nitro-2-xylol

OR6593-100G OR6593-25G OR6593-5G
APOSOR6593-100GEA 693 EUR
APOSOR6593-100G APOSOR6593-25G APOSOR6593-5G
2,3-Dimethyl-4-nitroanisol
2,3-Dimethyl-4-nitroanisol
Formel: C₉H₁₁NO₃
Schmelzpunkt: 70…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007163
CAS-Nummer: 81029-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR