2,3-Diaminobenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR6615-25G
APOSOR6615-25GEA
280
EUR
APOSOR6615-25G
2,3-Diaminobenzoesäure
2,3-Diaminobenzoic acid
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Schmelzpunkt: 178…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137818 CAS-Nummer: 603-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...