2-Aminoisonicotinonitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Aminopyridine-4-carbonitrile, 2-Amino-4-cyanopyridine
OR6619-1G
OR6619-5G
APOSOR6619-1GEA
21.9
EUR
APOSOR6619-1G
APOSOR6619-5G
2-Aminoisonicotinonitril
2-Aminoisonicotinonitril
Formel:
C₆H₅N₃ Molecular Weight: 119,13 g/mol Schmelzpunkt: 148…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791310 CAS-Nummer: 42182-27-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...