3-Chlor-2-pyridincarbonitril

Lieferant: Apollo Scientific
Achtung

OR6622-5G OR6622-10G
APOSOR6622-5GEA 21.9 EUR
APOSOR6622-5G APOSOR6622-10G
3-Chlor-2-pyridincarbonitril
3-Chlor-2-pyridincarbonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138.56 g/mol
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788834
CAS-Nummer: 38180-46-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR