3-Benzoylpropionsäure

Lieferant: Apollo Scientific

Synonyme: 4-Phenyl-4-oxobuttersäure, 4-Phenyl-4-ketobuttersäure

OR6625-100G
APOSOR6625-100GEA 94.4 EUR
APOSOR6625-100G
3-Benzoylpropionsäure
3-Benzoylpropionsäure
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002792
CAS-Nummer: 2051-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR