1-Brom-4-chlor-3-nitrobenzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-chloro-2-nitrobenzene, 5-Bromo-2-chloronitrobenzene
OR6626-100G
OR6626-25G
OR6626-5G
APOSOR6626-100GEA
71.2
EUR
APOSOR6626-100G
APOSOR6626-25G
APOSOR6626-5G
1-Brom-4-chlor-3-nitrobenzol
1-Bromo-4-chloro-3-nitrobenzene
Formel:
C₆H₃BrClNO₂ Molecular Weight: 236,4500 g/mol Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863216 CAS-Nummer: 16588-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...