2-(4-Methoxyphenyl)malondialdehyde 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(4-Methoxyphenyl)malonaldehyde

OR6632-1G OR6632-5G OR6632-25G OR6632-10G
APOSOR6632-1GEA 104 EUR
APOSOR6632-1G APOSOR6632-5G APOSOR6632-25G APOSOR6632-10G
2-(4-Methoxyphenyl)malondialdehyde 95+%
2-(4-Methoxyphenyl)malondialdehyde
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,187 g/mol
Schmelzpunkt: 145…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216504
CAS-Nummer: 65192-28-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR