3-Chlorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chlorphenyl)metaborsäure, Dihydroxy(3-chlorphenyl)boran, 3-Chlorbenzolboronsäure

OR6642-100G OR6642-500G
APOSOR6642-100GEA 81.2 EUR
APOSOR6642-100G APOSOR6642-500G
3-Chlorphenylboronsäure 98%
3-Chlorphenylboronsäure
Formel: C₆H₆BClO₂
Molecular Weight: 156,38 g/mol
Schmelzpunkt: 180…189 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00161354
CAS-Nummer: 63503-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR