2-Iodbenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR6669-1G
OR6669-5G
APOSOR6669-1GEA
29
EUR
APOSOR6669-1G
APOSOR6669-5G
2-Iodbenzaldehyd
2-Iodbenzaldehyd
Formel:
C₇H₅IO Molecular Weight: 232,02 g/mol Siedepunkt: 102 °C/3mmHg Schmelzpunkt: 36…37 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00039570 CAS-Nummer: 26260-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...