ω,3-Dichloracetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Chlorophenacyl chloride
OR6674-1G
OR6674-25G
OR6674-5G
APOSOR6674-1GEA
66.7
EUR
APOSOR6674-1G
APOSOR6674-25G
APOSOR6674-5G
ω,3-Dichloracetophenon
ω,3-Dichloracetophenon
Formel:
C₈H₆Cl₂O Schmelzpunkt: 40…42 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07783975 CAS-Nummer: 21886-56-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...