4-(Phenylsulphanyl)aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Phenylthio)aniline

OR6679-1G OR6679-5G OR6679-250MG
APOSOR6679-1GEA 92.8 EUR
APOSOR6679-1G APOSOR6679-5G APOSOR6679-250MG
4-(Phenylsulphanyl)aniline
4-(Phenylsulphanyl)aniline
Formel: C₁₂H₁₁NS
Lagertemperatur: Kühlschrank
MDL: MFCD00094027
CAS-Nummer: 1135-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR