5-Brom-2-chlorbenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR6702-100G OR6702-25G OR6702-5G
APOSOR6702-100GEA 88.6 EUR
APOSOR6702-100G APOSOR6702-25G APOSOR6702-5G
5-Brom-2-chlorbenzaldehyd
5-Brom-2-chlorbenzaldehyd
Formel: C₇H₄BrClO
Molecular Weight: 219,46 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445659
CAS-Nummer: 189628-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR