2-Brom-5-chlorbenzonitril

Lieferant: Apollo Scientific
Achtung

OR6705-25G OR6705-5G
APOSOR6705-25GEA 29 EUR
APOSOR6705-25G APOSOR6705-5G
2-Brom-5-chlorbenzonitril
2-Brom-5-chlorbenzonitril
Formel: C₇H₃BrClN
Molecular Weight: 216.46 g/mol
Schmelzpunkt: 138…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445660
CAS-Nummer: 57381-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR