3-Chlorisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloropyridine-4-carboxylic acid

OR6711-100G OR6711-25G OR6711-5G
APOSOR6711-100GEA 329 EUR
APOSOR6711-100G APOSOR6711-25G APOSOR6711-5G
3-Chlorisonicotinsäure
3-Chlorisonicotinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
Schmelzpunkt: 227…231 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03094685
CAS-Nummer: 88912-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR