4-Chlornicotinsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlor-3-pyridincarbonsäure, 4-Chlorpyridin-3-carbonsäure
OR6712-100G
OR6712-25G
OR6712-5G
APOSOR6712-100GEA
261
EUR
APOSOR6712-100G
APOSOR6712-25G
APOSOR6712-5G
4-Chlornicotinsäure 99%
4-Chlornicotinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: 139…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128860 CAS-Nummer: 10177-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...