4-(2-Methylthiazol-4-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR6745-1G OR6745-10G OR6745-5G
APOSOR6745-1GEA 149 EUR
APOSOR6745-1G APOSOR6745-10G APOSOR6745-5G
4-(2-Methylthiazol-4-yl)benzoic acid
4-(2-Methylthiazol-4-yl)benzoic acid
Formel: C₁₁H₉NO₂S
Molecular Weight: 219.26 g/mol
Schmelzpunkt: 263…268 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02225088
CAS-Nummer: 294620-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR