(4-(Benzyloxy)phenyl)hydrazine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6781-100G
OR6781-25G
OR6781-5G
APOSOR6781-100GEA
423
EUR
APOSOR6781-100G
APOSOR6781-25G
APOSOR6781-5G
(4-(Benzyloxy)phenyl)hydrazine hydrochloride 97%
(4-(Benzyloxy)phenyl)hydrazine hydrochloride
Formel:
C₁₃H₁₅ClN₂O Molecular Weight: 250,7240 g/mol Schmelzpunkt: 254 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00137741 CAS-Nummer: 52068-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...