1-Methyl-1H-indol-5-ylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-1-n-methylindole, 1-Methyl-1H-indol-5-amine, 5-Amino-1-methyl-1H-indole

OR6792-1G OR6792-500MG OR6792-5G
APOSOR6792-1GEA 29 EUR
APOSOR6792-1G APOSOR6792-500MG APOSOR6792-5G
1-Methyl-1H-indol-5-ylamine 97%
1-Methyl-1H-indol-5-ylamine
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Schmelzpunkt: 101…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03839873
CAS-Nummer: 102308-97-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR