3,4-(Methylendioxy)phenylessigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1,3-Benzodioxol-5-yl)acetic acid
OR6817-100G
OR6817-250G
OR6817-25G
APOSOR6817-100GEA
0
EUR
APOSOR6817-100G
APOSOR6817-250G
APOSOR6817-25G
3,4-(Methylendioxy)phenylessigsäure 98%
3,4-(Methylendioxy)phenylessigsäure
Formel:
C₉H₈O₄ Molecular Weight: 180,16 g/mol Schmelzpunkt: 125…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014576 CAS-Nummer: 2861-28-1 EINECS: 220-679-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...