2-Piperazin-1-ylphenol dihydrobromide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Hydroxyphenyl)piperazine dihydrobromide

OR6839-1G OR6839-5G
APOSOR6839-1GEA 21.9 EUR
APOSOR6839-1G APOSOR6839-5G
2-Piperazin-1-ylphenol dihydrobromide 98%
2-Piperazin-1-ylphenol dihydrobromide
Formel: C₁₀H₁₆Br₂N₂O
Schmelzpunkt: 260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040814
CAS-Nummer: 58260-69-8
EINECS: 261-190-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR